Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Octadien-1-ol, 95%

$363.27 - $363.27

Chemical Identifiers

Chemical Name or Material 2,4-Octadien-1-ol
CAS 56904-85-9
Molecular Formula CH3CH2CH2CH=CHCH=CHCH2OH
Formula Weight 126.21
MDL Number MFCD00014050
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1682006

 
alfa aesar™
A1682006
5g Each for $363.27
Description & Specifications

Specifications

Formula Weight 126.21
Chemical Name or Material 2,4-Octadien-1-ol
Assay 95%
Molecular Formula CH3CH2CH2CH=CHCH=CHCH2OH
MDL Number MFCD00014050
CAS 56904-85-9
Quantity 5g