missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Nonanedione, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0115114

 View more versions of this product

Catalog No. AAL0115114



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,4-Nonanedione
100°C to 103°C (19 mmHg)
1.4608
25 g
2,4-nonanedione, caproylacetone, 2,4-nonadione, acmc-20aobv
CCCCCC(=O)CC(=O)C
156.225
156.23
6175-23-1
C9H16O2
MFCD00015276
1757688
KFBXUKHERGLHLG-UHFFFAOYSA-N
nonane-2,4-dione
80314
≥98%

Chemical Identifiers

6175-23-1
156.225
KFBXUKHERGLHLG-UHFFFAOYSA-N
80314
CCCCCC(=O)CC(=O)C
C9H16O2
MFCD00015276
2,4-nonanedione, caproylacetone, 2,4-nonadione, acmc-20aobv
nonane-2,4-dione
Safety and Handling

Safety and Handling

EINECSNumber : 228-219-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only