missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Nonanedione, 98+%

Manufacturer:  Alfa Aesar™ L0115106

 View more versions of this product

Catalog No. AAL0115106



Specifications

Specifications

2,4-Nonanedione
100°C to 103°C (19mmHg)
6175-23-1
MFCD00015276
2,4-nonadione, 2,4-nonanedione, acmc-20aobv, caproylacetone
CCCCCC(=O)CC(=O)C
156.225
156.23
1.4608
5g
C9H16O2
1757688
KFBXUKHERGLHLG-UHFFFAOYSA-N
nonane-2,4-dione
80314
≥98%

Chemical Identifiers

6175-23-1
156.225
KFBXUKHERGLHLG-UHFFFAOYSA-N
80314
CCCCCC(=O)CC(=O)C
C9H16O2
MFCD00015276
2,4-nonadione, 2,4-nonanedione, acmc-20aobv, caproylacetone
nonane-2,4-dione
Safety and Handling

Safety and Handling

EINECSNumber : 228-219-9

TSCA : No

Recommended Storage : Ambient temperatures

Documents