missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Nitrophenyl)ethylamine hydrochloride, ≥98%

$32.53 - $467.26

Chemical Identifiers

CAS 29968-78-3
Molecular Formula C8H11ClN2O2
Molecular Weight (g/mol) 202.638
MDL Number MFCD00012900
InChI Key JVMHULJEYUQYSH-UHFFFAOYSA-N
Synonym 4-nitrophenethylamine hydrochloride, 4-nitrophenylethylamine hydrochloride, 4-nitrophenethylamine hcl, 2-4-nitrophenyl ethanamine hydrochloride, 2-4-nitrophenyl ethylamine hydrochloride, p-nitrophenylethylamine hcl, benzeneethanamine, 4-nitro-, monohydrochloride, 4-nitrophenethylaminehydrochloride, p-nitrophenethylamine, hydrochloride, pubchem11627
PubChem CID 458983
IUPAC Name 2-(4-nitrophenyl)ethanamine;hydrochloride
SMILES C1=CC(=CC=C1CCN)[N+](=O)[O-].Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6489403 View Documents Alfa Aesar™
H6489403
1g Each for $32.53
 
AAH6489406 View Documents Alfa Aesar™
H6489406
5g Each for $116.28
 
AAH6489414 View Documents Alfa Aesar™
H6489414
25g Each for $467.26
 
Description

Description

Applications
4-Nitrophenethylamine hydrochloride was used in the synthesis of ortho-metalated primary phenethylamines complexes containing six-membered palladacycles and N-(4-nitrophenethyl)formamide.

Solubility
Soluble in methanol almost transparency.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

29968-78-3
202.638
JVMHULJEYUQYSH-UHFFFAOYSA-N
458983
C1=CC(=CC=C1CCN)[N+](=O)[O-].Cl
C8H11ClN2O2
MFCD00012900
4-nitrophenethylamine hydrochloride, 4-nitrophenylethylamine hydrochloride, 4-nitrophenethylamine hcl, 2-4-nitrophenyl ethanamine hydrochloride, 2-4-nitrophenyl ethylamine hydrochloride, p-nitrophenylethylamine hcl, benzeneethanamine, 4-nitro-, monohydrochloride, 4-nitrophenethylaminehydrochloride, p-nitrophenethylamine, hydrochloride, pubchem11627
2-(4-nitrophenyl)ethanamine;hydrochloride

Specifications

Undesignated
1g
C8H11ClN2O2
3568915
JVMHULJEYUQYSH-UHFFFAOYSA-N
2-(4-nitrophenyl)ethanamine;hydrochloride
458983
Crystalline Powder
2-(4-Nitrophenyl)ethylamine hydrochloride
200°C (literature)
29968-78-3
MFCD00012900
4-nitrophenethylamine hydrochloride, 4-nitrophenylethylamine hydrochloride, 4-nitrophenethylamine hcl, 2-4-nitrophenyl ethanamine hydrochloride, 2-4-nitrophenyl ethylamine hydrochloride, p-nitrophenylethylamine hcl, benzeneethanamine, 4-nitro-, monohydrochloride, 4-nitrophenethylaminehydrochloride, p-nitrophenethylamine, hydrochloride, pubchem11627
C1=CC(=CC=C1CCN)[N+](=O)[O-].Cl
202.638
202.64
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning