missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Nitrophenyl)ethanol, 98%

Manufacturer:  Alfa Aesar™ H5450414

 View more versions of this product

Catalog No. AAH5450414



Description

Description

Applications
It is a phosphate protecting reagent and O6 of deoxyguanosine.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents. Keep at room temperature.
Specifications

Specifications

2-(4-Nitrophenyl)ethanol
59°C to 62°C
100-27-6
MFCD00010202
2-4-nitrophenyl ethan-1-ol
IKMXRUOZUUKSON-UHFFFAOYSA-N
2-(4-nitrophenyl)ethanol
7494
98%
177°C (16mmHg)
25g
C8H9NO3
1866148
Slightly soluble in water.
C1=CC(=CC=C1CCO)[N+](=O)[O-]
167.164
167.16

Chemical Identifiers

100-27-6
167.164
IKMXRUOZUUKSON-UHFFFAOYSA-N
7494
C1=CC(=CC=C1CCO)[N+](=O)[O-]
C8H9NO3
MFCD00010202
2-4-nitrophenyl ethan-1-ol
2-(4-nitrophenyl)ethanol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.

GHS P Statement
P280-P301+P310-P305+P351+P338-P321-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

EINECSNumber : 202-835-8

RTECSNumber : SG8602000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents