Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-n-Hexyloxyphenyl)-5-n-octylpyrimidine, 97%

$883.54 - $883.54

Chemical Identifiers

Chemical Name or Material 2-(4-n-Hexyloxyphenyl)-5-n-octylpyrimidine
CAS 57202-48-9
Molecular Formula C24H36N2O
Formula Weight 368.56
MDL Number MFCD00691439
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2260106

 
alfa aesar™
B2260106
5g Each for $883.54
Description & Specifications

Specifications

Formula Weight 368.56
Molecular Formula C24H36N2O
CAS 57202-48-9
Melting Point 57° to 62°C
Quantity 5g
Chemical Name or Material 2-(4-n-Hexyloxyphenyl)-5-n-octylpyrimidine
Assay 97%
MDL Number MFCD00691439