Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Morpholinyl)thiazole-5-carboxaldehyde, 97%

$113.01 - $388.74

Chemical Identifiers

Chemical Name or Material 2-(4-Morpholinyl)thiazole-5-carboxaldehyde
CAS 1011-41-2
Molecular Formula C8H10N2O2S
Formula Weight 198.25
MDL Number MFCD01568825
Synonym 2-(4-Morpholino)thiazole-5-carboxaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH50463MD

  SDS
alfa aesar™
H50463MD
250mg Each for $113.01

AAH5046303

  SDS
alfa aesar™
H5046303
1g Each for $388.74
Description & Specifications

Specifications

Formula Weight 198.25
Chemical Name or Material 2-(4-Morpholinyl)thiazole-5-carboxaldehyde
Assay 97%
Molecular Formula C8H10N2O2S
MDL Number MFCD01568825
CAS 1011-41-2
Synonym 2-(4-Morpholino)thiazole-5-carboxaldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 250mg
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.