Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Morpholinyl)pyridine, 96%

Chemical Identifiers

Chemical Name or Material 2-(4-Morpholinyl)pyridine
CAS 24255-25-2
Formula Weight 164.21
MDL Number MFCD01143321
Synonym 2-Morpholinopyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5598903

  SDS
alfa aesar™
H5598903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5598906

  SDS
alfa aesar™
H5598906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 164.21
Chemical Name or Material 2-(4-Morpholinyl)pyridine
Density 1.131g/mL
Physical Form Liquid
Assay 96%
MDL Number MFCD01143321
CAS 24255-25-2
Synonym 2-Morpholinopyridine
Boiling Point 135°C (20mmHg)
Quantity 1g