missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Morpholinyl)pyridine, 96%

Manufacturer:  Alfa Aesar™ H5598906

 View more versions of this product

Catalog No. AAH5598906



Specifications

Specifications

2-(4-Morpholinyl)pyridine
>110°C (230°F)
5g
C9H12N2O
2-4-morpholinyl pyridine
C1COCCN1C2=CC=CC=N2
164.208
164.21
96%
1.131 g/mL
135°C (20mmHg)
24255-25-2
MFCD01143321
XHPVOSNOIWGRQQ-UHFFFAOYSA-N
4-pyridin-2-ylmorpholine
10964853
Liquid

Chemical Identifiers

24255-25-2
164.208
XHPVOSNOIWGRQQ-UHFFFAOYSA-N
10964853
C1COCCN1C2=CC=CC=N2
C9H12N2O
MFCD01143321
2-4-morpholinyl pyridine
4-pyridin-2-ylmorpholine
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents