Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Morpholinyl)-5-(trifluoromethyl)aniline, 95%

Chemical Identifiers

Chemical Name or Material 2-(4-Morpholinyl)-5-(trifluoromethyl)aniline
CAS 784-57-6
Molecular Formula C11H13F3N2O
Formula Weight 246.23
MDL Number MFCD00042160
UN Number UN2811
Beilstein 213147
Synonym 3-Amino-4-(4-morpholino)benzotrifluoride; 4-[2-Amino-4-(trifluoromethyl)phenyl]morpholine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1264703

 
alfa aesar™
L1264703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1264706

 
alfa aesar™
L1264706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 246.23
Chemical Name or Material 2-(4-Morpholinyl)-5-(trifluoromethyl)aniline
Molecular Formula C11H13F3N2O
MDL Number MFCD00042160
CAS 784-57-6
UN Number UN2811
Synonym 3-Amino-4-(4-morpholino)benzotrifluoride; 4-[2-Amino-4-(trifluoromethyl)phenyl]morpholine
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 95%
Beilstein 213147
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 125° to 128°C
Quantity 1g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Applications
2-(4-Morpholinyl)-5-(trifluoromethyl)aniline is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.