missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Morpholinyl)-5-(trifluoromethyl)aniline, 95%

Manufacturer:  Alfa Aesar™ L1264706

 View more versions of this product

Catalog No. AAL1264706



Description

Description

Applications
2-(4-Morpholinyl)-5-(trifluoromethyl)aniline is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2-(4-Morpholinyl)-5-(trifluoromethyl)aniline
5g
C11H13F3N2O
UN2811
2-morpholino-5-trifluoromethyl aniline, 2-morpholin-4-yl-5-trifluoromethyl aniline, 3-amino-4-4-morpholino benzotrifluoride, 2-4-morpholinyl-5-trifluoromethyl aniline, n-2-amino-4-trifluoromethyl phenyl morpholine, 2-morpholin-4-yl-5-trifluoromethyl aniline, n-2-amino-4-trifluoromethylphenyl morpholine, n-2-amino-4-trifluoromethyl phenyl morpholine, 2-morpholino-5-trifluoromethylaniline, 3-amino-4-morpholino benzotrifluoride
C1COCCN1C2=C(C=C(C=C2)C(F)(F)F)N
246.233
246.23
125°C to 128°C
784-57-6
MFCD00042160
213147
CNVOJNRNRNAOOP-UHFFFAOYSA-N
2-morpholin-4-yl-5-(trifluoromethyl)aniline
136624
95%

Chemical Identifiers

784-57-6
246.233
CNVOJNRNRNAOOP-UHFFFAOYSA-N
136624
C1COCCN1C2=C(C=C(C=C2)C(F)(F)F)N
C11H13F3N2O
MFCD00042160
2-morpholino-5-trifluoromethyl aniline, 2-morpholin-4-yl-5-trifluoromethyl aniline, 3-amino-4-4-morpholino benzotrifluoride, 2-4-morpholinyl-5-trifluoromethyl aniline, n-2-amino-4-trifluoromethyl phenyl morpholine, 2-morpholin-4-yl-5-trifluoromethyl aniline, n-2-amino-4-trifluoromethylphenyl morpholine, n-2-amino-4-trifluoromethyl phenyl morpholine, 2-morpholino-5-trifluoromethylaniline, 3-amino-4-morpholino benzotrifluoride
2-morpholin-4-yl-5-(trifluoromethyl)aniline
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents