missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[2-(4-Methylpiperazin-1-yl)phenyl]methanol, 97%, Maybridge

Chemical Identifiers

CAS 123987-12-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C12H18N2O
Formula Weight 206.29
MDL Number MFCD03407383
Chemical Name or Material [2-(4-Methylpiperazin-1-yl)phenyl]methanol
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC36009CB SDS Fisher Scientific Company
CC36009CB
Amber glass bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC36009EA SDS Fisher Scientific Company
CC36009EA
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC36009DA SDS Fisher Scientific Company
CC36009DA
Amber glass bottle 1mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

123987-12-2
100.0
C12H18N2O
MFCD03407383
97%
96.5
97%
206.29
[2-(4-Methylpiperazin-1-yl)phenyl]methanol

Specifications

250mg
96.5
Amber glass bottle
C12H18N2O
MFCD03407383
97%
123987-12-2
100.0
97%
206.29
[2-(4-Methylpiperazin-1-yl)phenyl]methanol

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger