Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Methylphenyl)thioacetamide, 90+%

$99.89 - $245.61

Chemical Identifiers

Chemical Name or Material 2-(4-Methylphenyl)thioacetamide
CAS 97426-53-4
Molecular Formula C9H11NS
Formula Weight 165.25
MDL Number MFCD00278201
Synonym 2-(p-Tolyl)thioacetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2627803

  SDS
alfa aesar™
H2627803
1g Each for $245.61

AAH26278MD

  SDS
alfa aesar™
H26278MD
250mg Each for $99.89
Description & Specifications

Specifications

Formula Weight 165.25
Assay ≥90%
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 97426-53-4
Chemical Name or Material 2-(4-Methylphenyl)thioacetamide
Melting Point 111° to 114°C
Molecular Formula C9H11NS
MDL Number MFCD00278201
Synonym 2-(p-Tolyl)thioacetamide
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.