missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Methylphenyl)thiazole-4-carboxylic acid, 97%

$38.74 - $506.04

Chemical Identifiers

CAS 17228-99-8
Molecular Formula C11H9NO2S
Molecular Weight (g/mol) 219.258
MDL Number MFCD01936002
InChI Key MTVWIZYQYPRZGZ-UHFFFAOYSA-N
Synonym 2-4-methylphenyl thiazole-4-carboxylic acid
PubChem CID 2794795
IUPAC Name 2-(4-methylphenyl)-1,3-thiazole-4-carboxylic acid
SMILES CC1=CC=C(C=C1)C2=NC(=CS2)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54054MD View Documents Alfa Aesar™
H54054MD
250mg Each for $38.74
 
AAH5405403 View Documents Alfa Aesar™
H5405403
1g Each for $122.31
 
AAH5405406 View Documents Alfa Aesar™
H5405406
5g Each for $506.04
 
Description

Description

Applications
Used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

17228-99-8
219.258
MTVWIZYQYPRZGZ-UHFFFAOYSA-N
2794795
CC1=CC=C(C=C1)C2=NC(=CS2)C(=O)O
C11H9NO2S
MFCD01936002
2-4-methylphenyl thiazole-4-carboxylic acid
2-(4-methylphenyl)-1,3-thiazole-4-carboxylic acid

Specifications

168°C to 172°C
17228-99-8
MFCD01936002
Slightly soluble in water.
CC1=CC=C(C=C1)C2=NC(=CS2)C(=O)O
219.258
219.26
2-(4-Methylphenyl)thiazole-4-carboxylic acid
250mg
C11H9NO2S
2-4-methylphenyl thiazole-4-carboxylic acid
MTVWIZYQYPRZGZ-UHFFFAOYSA-N
2-(4-methylphenyl)-1,3-thiazole-4-carboxylic acid
2794795
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures