missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Methylphenyl)thiazole-4-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H54054MD

 View more versions of this product

Catalog No. AAH54054MD



Description

Description

Applications
Used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

2-(4-Methylphenyl)thiazole-4-carboxylic acid
250mg
C11H9NO2S
2-4-methylphenyl thiazole-4-carboxylic acid
CC1=CC=C(C=C1)C2=NC(=CS2)C(=O)O
219.258
219.26
168°C to 172°C
17228-99-8
MFCD01936002
MTVWIZYQYPRZGZ-UHFFFAOYSA-N
2-(4-methylphenyl)-1,3-thiazole-4-carboxylic acid
2794795
97%

Chemical Identifiers

17228-99-8
219.258
MTVWIZYQYPRZGZ-UHFFFAOYSA-N
2794795
CC1=CC=C(C=C1)C2=NC(=CS2)C(=O)O
C11H9NO2S
MFCD01936002
2-4-methylphenyl thiazole-4-carboxylic acid
2-(4-methylphenyl)-1,3-thiazole-4-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents