missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Methylphenyl)imidazole-4,5-dithiocarboxamide, 97%

Chemical Identifiers

CAS 1347815-08-0
Molecular Formula C12H12N4S2
Molecular Weight (g/mol) 276.376
MDL Number MFCD20265364
InChI Key CFNPTMBECAYGCK-UHFFFAOYSA-N
Synonym 2-4-methylphenyl imidazole-4,5-dithiocarboxamide
PubChem CID 73996015
IUPAC Name 2-(4-methylphenyl)-1H-imidazole-4,5-dicarbothioamide
SMILES CC1=CC=C(C=C1)C2=NC(=C(N2)C(=S)N)C(=S)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54891MD View Documents Alfa Aesar™
H54891MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5489103 View Documents Alfa Aesar™
H5489103
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5489106 View Documents Alfa Aesar™
H5489106
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1347815-08-0
276.376
CFNPTMBECAYGCK-UHFFFAOYSA-N
73996015
CC1=CC=C(C=C1)C2=NC(=C(N2)C(=S)N)C(=S)N
C12H12N4S2
MFCD20265364
2-4-methylphenyl imidazole-4,5-dithiocarboxamide
2-(4-methylphenyl)-1H-imidazole-4,5-dicarbothioamide

Specifications

250mg
C12H12N4S2
2-4-methylphenyl imidazole-4,5-dithiocarboxamide
CC1=CC=C(C=C1)C2=NC(=C(N2)C(=S)N)C(=S)N
276.376
276.37
2-(4-Methylphenyl)imidazole-4,5-dithiocarboxamide
1347815-08-0
MFCD20265364
CFNPTMBECAYGCK-UHFFFAOYSA-N
2-(4-methylphenyl)-1H-imidazole-4,5-dicarbothioamide
73996015
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures