missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Methylphenyl)ethanol, 99%

$47.73 - $151.09

Chemical Identifiers

CAS 699-02-5
Molecular Formula C9H12O
Formula Weight 136.2
MDL Number MFCD00002904
Synonym 4-Methylphenethyl alcohol; 2-(p-Tolyl)ethanol
Chemical Name or Material 2-(4-Methylphenyl)ethanol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2091803 SDS Alfa Aesar™
B2091803
1g Each for $47.73
AAB2091806 SDS Alfa Aesar™
B2091806
5g Each for $151.09
AAB2091814 SDS Alfa Aesar™
B2091814
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

699-02-5
136.2
4-Methylphenethyl alcohol; 2-(p-Tolyl)ethanol
C9H12O
MFCD00002904
2-(4-Methylphenyl)ethanol

Specifications

699-02-5
136.2
4-Methylphenethyl alcohol; 2-(p-Tolyl)ethanol
1g
2-(4-Methylphenyl)ethanol
C9H12O
MFCD00002904
244° to 245°C
107°C (225°F)
99%

Safety and Handling

TSCA : TSCA