missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Methylphenyl)acetamidoxime, 97%

Chemical Identifiers

CAS 1312766-67-8
Molecular Formula C9H12N2O
Molecular Weight (g/mol) 164.208
InChI Key CKMSCSXWLFGEGO-UHFFFAOYSA-N
Synonym n-hydroxy-2-p-tolyl-acetamidine, 2-p-tolyl-acetamidoxime, 2-4-tolyl-acetamidoxime, z-n'-hydroxy-2-p-tolyl acetimidamide
PubChem CID 9679732
IUPAC Name N'-hydroxy-2-(4-methylphenyl)ethanimidamide
SMILES CC1=CC=C(C=C1)CC(=NO)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66026MD View Documents Alfa Aesar™
H66026MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6602603 View Documents Alfa Aesar™
H6602603
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1312766-67-8
164.208
n-hydroxy-2-p-tolyl-acetamidine, 2-p-tolyl-acetamidoxime, 2-4-tolyl-acetamidoxime, z-n'-hydroxy-2-p-tolyl acetimidamide
N'-hydroxy-2-(4-methylphenyl)ethanimidamide
C9H12N2O
CKMSCSXWLFGEGO-UHFFFAOYSA-N
9679732
CC1=CC=C(C=C1)CC(=NO)N

Specifications

Cream or White
1312766-67-8
n-hydroxy-2-p-tolyl-acetamidine, 2-p-tolyl-acetamidoxime, 2-4-tolyl-acetamidoxime, z-n'-hydroxy-2-p-tolyl acetimidamide
CC1=CC=C(C=C1)CC(=NO)N
164.208
164.2
97%
250mg
C9H12N2O
CKMSCSXWLFGEGO-UHFFFAOYSA-N
N'-hydroxy-2-(4-methylphenyl)ethanimidamide
9679732
Crystalline Solid
2-(4-Methylphenyl)acetamidoxime