Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Methylphenyl)-4,5-diphenylthiazole, 97%

$253.46 - $253.46

Chemical Identifiers

Chemical Name or Material 2-(4-Methylphenyl)-4,5-diphenylthiazole
CAS 120332-29-8
Molecular Formula C22H17NS
Formula Weight 327.44g/mol
MDL Number MFCD16251583
Synonym 4,5-Diphenyl-2-(p-tolyl)thiazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH51847MD

 
alfa aesar™
H51847-MD
Each for $253.46
Description & Specifications

Specifications

Melting Point 78° to 83°C
Quantity 250mg
Chemical Name or Material 2-(4-Methylphenyl)-4,5-diphenylthiazole
CAS 120332-29-8
Assay 97%
Molecular Formula C22H17NS
Formula Weight 327.44g/mol
MDL Number MFCD16251583
Synonym 4,5-Diphenyl-2-(p-tolyl)thiazole
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.