Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Methylphenoxy)nicotinic acid, 96%

$203.03 - $203.03

Chemical Identifiers

Chemical Name or Material 2-(4-Methylphenoxy)nicotinic acid
CAS 54629-15-1
Molecular Formula C13H11NO3
Formula Weight 229.24g/mol
MDL Number MFCD00052118
Synonym 2-(p-Tolyloxy)pyridine-3-carboxylic acid; 2-(p-Tolyloxy)nicotinic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5028903

 
alfa aesar™
H50289-03
1g Each for $203.03
Description & Specifications

Specifications

Quantity 1g
Quantity 1g
Chemical Name or Material 2-(4-Methylphenoxy)nicotinic acid
CAS 54629-15-1
Assay 96%
Molecular Formula C13H11NO3
Formula Weight 229.24g/mol
MDL Number MFCD00052118
Synonym 2-(p-Tolyloxy)pyridine-3-carboxylic acid; 2-(p-Tolyloxy)nicotinic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.