Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Methylphenoxy)benzaldehyde, 97%

$236.01 - $236.01

Chemical Identifiers

Chemical Name or Material 2-(4-Methylphenoxy)benzaldehyde
CAS 19434-35-6
Molecular Formula C14H12O2
Formula Weight 212.25g/mol
MDL Number MFCD01568738
Synonym 2-(p-Tolyloxy)benzaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5046103

 
alfa aesar™
H50461-03
Each for $236.01
Description & Specifications

Specifications

Melting Point 55°C
Quantity 1g
Chemical Name or Material 2-(4-Methylphenoxy)benzaldehyde
CAS 19434-35-6
Assay 97%
Molecular Formula C14H12O2
Formula Weight 212.25g/mol
MDL Number MFCD01568738
Synonym 2-(p-Tolyloxy)benzaldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.