missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Methylperhydro-1,4-diazepin-1-yl)benzylamine, 90%, Maybridge

Chemical Identifiers

CAS 915707-56-1
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Molecular Formula C13H21N3
Formula Weight 219.33
MDL Number MFCD09065001
Chemical Name or Material 2-(4-Methylperhydro-1, 4-diazepin-1-yl)benzylamine
Assay 90%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC60413CB SDS Fisher Scientific Company
CC60413CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC60413DA SDS Fisher Scientific Company
CC60413DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

915707-56-1
100.0
C13H21N3
MFCD09065001
90%
89.5
90%
219.33
2-(4-Methylperhydro-1, 4-diazepin-1-yl)benzylamine

Specifications

250mg
89.5
Amber Glass Bottle
C13H21N3
MFCD09065001
90%
915707-56-1
100.0
90%
219.33
2-(4-Methylperhydro-1, 4-diazepin-1-yl)benzylamine

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger