missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Methylperhydro-1,4-diazepin-1-yl)benzonitrile, 95%, Maybridge

Chemical Identifiers

CAS 204078-93-3
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C13H17N3
Formula Weight 215.3
MDL Number MFCD09065002
Chemical Name or Material 2-(4-Methylperhydro-1, 4-diazepin-1-yl)benzonitrile
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC60416CB SDS Fisher Scientific Company
CC60416CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

204078-93-3
100.0
C13H17N3
MFCD09065002
95%
94.5
95%
215.3
2-(4-Methylperhydro-1, 4-diazepin-1-yl)benzonitrile

Specifications

215.3
100.0
MFCD09065002
94.5
204078-93-3
95%
250mg
95%
C13H17N3
Amber Glass Bottle
2-(4-Methylperhydro-1, 4-diazepin-1-yl)benzonitrile

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger