Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Methylbenzylamino)pyridine-5-boronic acid pinacol ester, 95%

$210.95 - $479.15

Chemical Identifiers

Chemical Name or Material 2-(4-Methylbenzylamino)pyridine-5-boronic acid pinacol ester
CAS 1073354-32-1
Molecular Formula C19H25BN2O2
Formula Weight 324.23
MDL Number MFCD06798272
Synonym N-(4-Methylbenzyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27434MD

  SDS
alfa aesar™
H27434MD
250mg Each for $210.95

AAH2743403

  SDS
alfa aesar™
H2743403
1g Each for $479.15
Description & Specifications

Specifications

Chemical Name or Material 2-(4-Methylbenzylamino)pyridine-5-boronic acid pinacol ester
CAS 1073354-32-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 95%
Molecular Formula C19H25BN2O2
Formula Weight 324.23
MDL Number MFCD06798272
Synonym N-(4-Methylbenzyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridinamine
Melting Point 148° to 153°C
Quantity 250mg
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.