missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Methyl-5-thiazolyl)ethyl Hexanoate 98.0+%, TCI America™

Manufacturer:  Tci America M249125G

 View more versions of this product

Catalog No. M249125G



Specifications

Specifications

2-(4-Methyl-5-thiazolyl)ethyl Hexanoate
Undesignated
94159-32-7
MFCD09032914
VJULDCZELAIZHC-UHFFFAOYSA-N
2-(4-methyl-1,3-thiazol-5-yl)ethyl hexanoate
3023839
Clear Liquid at 20°C
152°C
25g
C12H19NO2S
Hexanoic Acid 2-(4-Methyl-5-thiazolyl)ethyl Ester, 5-(2-Hexanoyloxyethyl)-4-methylthiazole
CCCCCC(=O)OCCC1=C(N=CS1)C
241.349
241.35
≥98.0% (GC)

Chemical Identifiers

94159-32-7
241.349
VJULDCZELAIZHC-UHFFFAOYSA-N
3023839
CCCCCC(=O)OCCC1=C(N=CS1)C
C12H19NO2S
MFCD09032914
Hexanoic Acid 2-(4-Methyl-5-thiazolyl)ethyl Ester, 5-(2-Hexanoyloxyethyl)-4-methylthiazole
2-(4-methyl-1,3-thiazol-5-yl)ethyl hexanoate
Safety and Handling

Safety and Handling

TSCA : No

Documents
Promotions

Promotions