missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(4-Methyl-5-thiazolyl)ethyl acetate, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2186722
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-(4-Methyl-5-thiazolyl)ethyl acetate | |
1.147 | |
>110°C (230°F) | |
1.51 | |
100 g | |
CRTCWNPLKVVXIX-UHFFFAOYSA-N | |
2-(4-methyl-1,3-thiazol-5-yl)ethyl acetate | |
61192 | |
≥98% |
656-53-1 | |
116°C to 118°C (6 mmHg) | |
C8H11NO2S | |
MFCD00005338 | |
2-4-methylthiazol-5-yl ethyl acetate, 4-methyl-5-thiazolylethyl acetate, 2-4-methyl-5-thiazolyl ethyl acetate, sulfurol acetate, 4-methyl-5-2-acetoxyethyl thiazole, 4-methyl-5-thiazoleethanol acetate, 5-2-acetoxyethyl-4-methylthiazole, 2-4-methyl-1,3-thiazol-5-yl ethyl acetate, 5-thiazoleethanol, 4-methyl-, acetate ester, 4-methyl-5-beta-acetoxyethyl thiazole | |
CC(=O)OCCC1=C(C)N=CS1 | |
185.24 | |
185.24 |
Chemical Identifiers
656-53-1 | |
185.24 | |
CRTCWNPLKVVXIX-UHFFFAOYSA-N | |
61192 | |
CC(=O)OCCC1=C(C)N=CS1 |
C8H11NO2S | |
MFCD00005338 | |
2-4-methylthiazol-5-yl ethyl acetate, 4-methyl-5-thiazolylethyl acetate, 2-4-methyl-5-thiazolyl ethyl acetate, sulfurol acetate, 4-methyl-5-2-acetoxyethyl thiazole, 4-methyl-5-thiazoleethanol acetate, 5-2-acetoxyethyl-4-methylthiazole, 2-4-methyl-1,3-thiazol-5-yl ethyl acetate, 5-thiazoleethanol, 4-methyl-, acetate ester, 4-methyl-5-beta-acetoxyethyl thiazole | |
2-(4-methyl-1,3-thiazol-5-yl)ethyl acetate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 211-515-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only