Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Methyl-1-piperazinyl)thiazole-5-carboxaldehyde, 99%

$114.51 - $396.28

Chemical Identifiers

Chemical Name or Material 2-(4-Methyl-1-piperazinyl)thiazole-5-carboxaldehyde
CAS 320423-50-5
Molecular Formula C9H13N3OS
Formula Weight 211.29
MDL Number MFCD01568852
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5046603

  SDS
alfa aesar™
H5046603
1g Each for $396.28

AAH50466MD

  SDS
alfa aesar™
H50466MD
250mg Each for $114.51
Description & Specifications

Specifications

Formula Weight 211.29
CAS 320423-50-5
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 99%
Molecular Formula C9H13N3OS
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Chemical Name or Material 2-(4-Methyl-1-piperazinyl)thiazole-5-carboxaldehyde
MDL Number MFCD01568852
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.