missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Methoxystyryl)-4,6-bis(trichloromethyl)-1,3,5-triazine, TCI America™

$135.97 - $447.64

Chemical Identifiers

CAS 42573-57-9
Molecular Formula C14H9Cl6N3O
Molecular Weight (g/mol) 447.946
MDL Number MFCD01940892
InChI Key MCNPOZMLKGDJGP-QPJJXVBHSA-N
PubChem CID 6014381
IUPAC Name 2-[(E)-2-(4-methoxyphenyl)ethenyl]-4,6-bis(trichloromethyl)-1,3,5-triazine
SMILES COC1=CC=C(C=C1)C=CC2=NC(=NC(=N2)C(Cl)(Cl)Cl)C(Cl)(Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M214025G View Documents TCI America
M214025G
25g Each for $447.64
 
M21405G View Documents TCI America
M21405G
5g Each for $135.97
 
Specifications

Chemical Identifiers

42573-57-9
447.946
MCNPOZMLKGDJGP-QPJJXVBHSA-N
2-[(E)-2-(4-methoxyphenyl)ethenyl]-4,6-bis(trichloromethyl)-1,3,5-triazine
C14H9Cl6N3O
MFCD01940892
6014381
COC1=CC=C(C=C1)C=CC2=NC(=NC(=N2)C(Cl)(Cl)Cl)C(Cl)(Cl)Cl

Specifications

Yellow
25g
C14H9Cl6N3O
MCNPOZMLKGDJGP-QPJJXVBHSA-N
2-[(E)-2-(4-methoxyphenyl)ethenyl]-4,6-bis(trichloromethyl)-1,3,5-triazine
6014381
Crystal-Powder at 20°C
2-(4-Methoxystyryl)-4,6-bis(trichloromethyl)-1,3,5-triazine
193°C
42573-57-9
MFCD01940892
COC1=CC=C(C=C1)C=CC2=NC(=NC(=N2)C(Cl)(Cl)Cl)C(Cl)(Cl)Cl
447.946
447.95
≥98.0% (HPLC,N)
Safety and Handling

Safety and Handling

TSCA : Yes