Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Methoxyphenyl)thioacetamide, 97%

$142.07 - $423.30

Chemical Identifiers

Chemical Name or Material 2-(4-Methoxyphenyl)thioacetamide
CAS 60759-02-6
Molecular Formula C9H11NOS
Formula Weight 181.27
MDL Number MFCD00278203
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2674203

 
alfa aesar™
H2674203
1g Each for $142.07

AAH2674206

 
alfa aesar™
H2674206
5g Each for $423.30
Description & Specifications

Specifications

Formula Weight 181.27
Molecular Formula C9H11NOS
Chemical Name or Material 2-(4-Methoxyphenyl)thioacetamide
Assay 97%
MDL Number MFCD00278203
CAS 60759-02-6
Melting Point 139° to 142°C
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
They find extensive use in organic synthesis. In pharmaceuticals extensively used for instance as antibacterials.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.