missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Methoxyphenyl)pyrrolidine, TCI America™

$189.45 - $661.55

Chemical Identifiers

CAS 74190-66-2
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD01631836
InChI Key HCBLZPKEGFYHDN-UHFFFAOYSA-N
Synonym 4-(2-Pyrrolidinyl)anisole
PubChem CID 3756957
IUPAC Name 2-(4-methoxyphenyl)pyrrolidine
SMILES COC1=CC=C(C=C1)C2CCCN2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M24461G View Documents TCI America
M24461G
1g Each for $661.55
 
M2446200MG View Documents TCI America
M2446200MG
200mg Each for $189.45
 
Specifications

Chemical Identifiers

74190-66-2
177.247
HCBLZPKEGFYHDN-UHFFFAOYSA-N
3756957
COC1=CC=C(C=C1)C2CCCN2
C11H15NO
MFCD01631836
4-(2-Pyrrolidinyl)anisole
2-(4-methoxyphenyl)pyrrolidine

Specifications

112°C
1g
C11H15NO
4-(2-Pyrrolidinyl)anisole
COC1=CC=C(C=C1)C2CCCN2
177.247
177.25
≥98.0% (T)
Yellow
74190-66-2
MFCD01631836
HCBLZPKEGFYHDN-UHFFFAOYSA-N
2-(4-methoxyphenyl)pyrrolidine
3756957
Clear Liquid at 20°C
2-(4-Methoxyphenyl)pyrrolidine
Safety and Handling

Safety and Handling

TSCA : No