missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Methoxyphenyl)pyrrolidine 98.0+%, TCI America™

Manufacturer:  Tci America M2446200MG

 View more versions of this product

Catalog No. M2446200MG



Specifications

Specifications

2-(4-Methoxyphenyl)pyrrolidine
Yellow
74190-66-2
MFCD01631836
HCBLZPKEGFYHDN-UHFFFAOYSA-N
2-(4-methoxyphenyl)pyrrolidine
3756957
Clear Liquid at 20°C
112°C
200mg
C11H15NO
4-(2-Pyrrolidinyl)anisole
COC1=CC=C(C=C1)C2CCCN2
177.247
177.25
≥98.0% (T)

Chemical Identifiers

74190-66-2
177.247
HCBLZPKEGFYHDN-UHFFFAOYSA-N
3756957
COC1=CC=C(C=C1)C2CCCN2
C11H15NO
MFCD01631836
4-(2-Pyrrolidinyl)anisole
2-(4-methoxyphenyl)pyrrolidine
Safety and Handling

Safety and Handling

TSCA : No

Documents
Promotions

Promotions