missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Methoxyphenyl)indole, TCI America™

$220.00 - $660.02

Chemical Identifiers

CAS 5784-95-2
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.275
MDL Number MFCD00443483
InChI Key BHCBPEBRFMLOND-UHFFFAOYSA-N
PubChem CID 231244
IUPAC Name 2-(4-methoxyphenyl)-1H-indole
SMILES COC1=CC=C(C=C1)C2=CC3=CC=CC=C3N2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M27891G View Documents TCI America
M27891G
1g Each for $220.00
 
M27895G View Documents TCI America
M27895G
5g Each for $660.02
 
Specifications

Chemical Identifiers

5784-95-2
223.275
BHCBPEBRFMLOND-UHFFFAOYSA-N
2-(4-methoxyphenyl)-1H-indole
C15H13NO
MFCD00443483
231244
COC1=CC=C(C=C1)C2=CC3=CC=CC=C3N2

Specifications

Undesignated
1g
C15H13NO
BHCBPEBRFMLOND-UHFFFAOYSA-N
2-(4-methoxyphenyl)-1H-indole
231244
Crystal-Powder at 20°C
2-(4-Methoxyphenyl)indole
231°C
5784-95-2
MFCD00443483
COC1=CC=C(C=C1)C2=CC3=CC=CC=C3N2
223.275
223.28
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No