missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Methoxyphenyl)ethylamine, TCI America™

Manufacturer:  TCI America M0795250ML

 View more versions of this product

Catalog No. M0795250ML



Specifications

Specifications

2-(4-Methoxyphenyl)ethylamine
Yellow
55-81-2
MFCD00008192
4-(2-Aminoethyl)anisole, 4-Methoxyphenethylamine
COC1=CC=C(C=C1)CCN
151.209
CHEBI:266039
Clear Liquid at 20°C
138°C
250mL
C9H13NO
2735
LTPVSOCPYWDIFU-UHFFFAOYSA-N
2-(4-methoxyphenyl)ethanamine
4657
151.21
≥98.0% (GC,T)

Chemical Identifiers

55-81-2
151.209
LTPVSOCPYWDIFU-UHFFFAOYSA-N
4657
2-(4-methoxyphenyl)ethanamine
C9H13NO
MFCD00008192
4-(2-Aminoethyl)anisole, 4-Methoxyphenethylamine
CHEBI:266039
COC1=CC=C(C=C1)CCN
Safety and Handling

Safety and Handling

RTECSNumber : SH7875000

TSCA : Yes

Documents