missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Methoxyphenyl)acetamidoxime, 97%

Chemical Identifiers

CAS 1055926-31-2
Molecular Formula C9H12N2O2
Molecular Weight (g/mol) 180.207
InChI Key QIACLRIKXICINP-UHFFFAOYSA-N
Synonym n'-hydroxy-2-4-methoxyphenyl ethanimidamide, 1z-n'-hydroxy-2-4-methoxyphenyl ethanimidamide, n-hydroxy-2-4-methoxy-phenyl-acetamidine, z-n'-hydroxy-2-4-methoxyphenyl ethanimidamide, 2-4-methoxyphenyl-acetamidoxime, benzeneethanimidamide, n-hydroxy-4-methoxy, z-n'-hydroxy-2-4-methoxyphenyl acetimidamide, benzeneethanimidamide,n-hydroxy-4-methoxy, 1z-n\'-hydroxy-2-4-methoxyphenyl ethanimidamide
PubChem CID 5356867
IUPAC Name N'-hydroxy-2-(4-methoxyphenyl)ethanimidamide
SMILES COC1=CC=C(C=C1)CC(=NO)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66318MD View Documents Alfa Aesar™
H66318MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6631803 View Documents Alfa Aesar™
H6631803
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1055926-31-2
180.207
n'-hydroxy-2-4-methoxyphenyl ethanimidamide, 1z-n'-hydroxy-2-4-methoxyphenyl ethanimidamide, n-hydroxy-2-4-methoxy-phenyl-acetamidine, z-n'-hydroxy-2-4-methoxyphenyl ethanimidamide, 2-4-methoxyphenyl-acetamidoxime, benzeneethanimidamide, n-hydroxy-4-methoxy, z-n'-hydroxy-2-4-methoxyphenyl acetimidamide, benzeneethanimidamide,n-hydroxy-4-methoxy, 1z-n\'-hydroxy-2-4-methoxyphenyl ethanimidamide
N'-hydroxy-2-(4-methoxyphenyl)ethanimidamide
C9H12N2O2
QIACLRIKXICINP-UHFFFAOYSA-N
5356867
COC1=CC=C(C=C1)CC(=NO)N

Specifications

White
1055926-31-2
n'-hydroxy-2-4-methoxyphenyl ethanimidamide, 1z-n'-hydroxy-2-4-methoxyphenyl ethanimidamide, n-hydroxy-2-4-methoxy-phenyl-acetamidine, z-n'-hydroxy-2-4-methoxyphenyl ethanimidamide, 2-4-methoxyphenyl-acetamidoxime, benzeneethanimidamide, n-hydroxy-4-methoxy, z-n'-hydroxy-2-4-methoxyphenyl acetimidamide, benzeneethanimidamide,n-hydroxy-4-methoxy, 1z-n\'-hydroxy-2-4-methoxyphenyl ethanimidamide
COC1=CC=C(C=C1)CC(=NO)N
180.207
180.2
97%
250mg
C9H12N2O2
QIACLRIKXICINP-UHFFFAOYSA-N
N'-hydroxy-2-(4-methoxyphenyl)ethanimidamide
5356867
Solid
2-(4-Methoxyphenyl)acetamidoxime