missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Methoxyphenyl)-4-methylthiazole-5-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H5403806

 View more versions of this product

Catalog No. AAH5403806



Specifications

Specifications

2-(4-Methoxyphenyl)-4-methylthiazole-5-carboxylic acid
5g
C12H11NO3S
2-4-methoxy phenyl-4-methyl-1,3-thiazol-5-carboxylic acid
CC1=C(SC(=N1)C2=CC=C(C=C2)OC)C(=O)O
249.284
249.28
236°C to 241°C
54001-16-0
MFCD06857928
HWHHZTHVYFDJNK-UHFFFAOYSA-N
2-(4-methoxyphenyl)-4-methyl-1,3-thiazole-5-carboxylic acid
4962843
97%

Chemical Identifiers

54001-16-0
249.284
HWHHZTHVYFDJNK-UHFFFAOYSA-N
4962843
CC1=C(SC(=N1)C2=CC=C(C=C2)OC)C(=O)O
C12H11NO3S
MFCD06857928
2-4-methoxy phenyl-4-methyl-1,3-thiazol-5-carboxylic acid
2-(4-methoxyphenyl)-4-methyl-1,3-thiazole-5-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents