Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Methoxyphenoxy)propylamine, 99%

Chemical Identifiers

Chemical Name or Material 2-(4-Methoxyphenoxy)propylamine
CAS 93750-30-2
Molecular Formula C10H15NO2
Formula Weight 181.24
MDL Number MFCD02684116
UN Number UN2735
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5072103

  SDS
alfa aesar™
H5072103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH50721MD

  SDS
alfa aesar™
H50721MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 181.24
Quantity 1g
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
MDL Number MFCD02684116
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
CAS 93750-30-2
Health Hazard 1 Danger
Chemical Name or Material 2-(4-Methoxyphenoxy)propylamine
Assay 99%
UN Number UN2735
Molecular Formula C10H15NO2
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.