missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Methoxyphenoxy)ethylamine, Maybridge™

Chemical Identifiers

CAS 50800-92-5
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.208
MDL Number MFCD00052978
InChI Key YXRCDWIZAGWUBL-UHFFFAOYSA-N
Synonym 1-2-aminoethoxy-4-methoxybenzene, 2-4-methoxy-phenoxy ethylamine, 2-4-methoxy-phenoxy-ethylamine, 2-4-methoxyphenoxy ethan-1-amine, 2-4-methoxyphenoxy ethanamine, 2-4-methoxyphenoxy ethylamine, 2-4-methoxyphenoxy-1-ethanamine, ethanamine, 2-4-methoxyphenoxy, maybridge3_000601
PubChem CID 1797793
IUPAC Name 2-(4-methoxyphenoxy)ethanamine
SMILES COC1=CC=C(C=C1)OCCN
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
RF03346CB
SDS View Documents
Fisher Scientific Company
RF03346CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

50800-92-5
167.208
YXRCDWIZAGWUBL-UHFFFAOYSA-N
1797793
COC1=CC=C(C=C1)OCCN
C9H13NO2
MFCD00052978
1-2-aminoethoxy-4-methoxybenzene, 2-4-methoxy-phenoxy ethylamine, 2-4-methoxy-phenoxy-ethylamine, 2-4-methoxyphenoxy ethan-1-amine, 2-4-methoxyphenoxy ethanamine, 2-4-methoxyphenoxy ethylamine, 2-4-methoxyphenoxy-1-ethanamine, ethanamine, 2-4-methoxyphenoxy, maybridge3_000601
2-(4-methoxyphenoxy)ethanamine

Specifications

Amber Glass Bottle
50800-92-5
NH2(CH2)2OC6H4OCH3
1-2-aminoethoxy-4-methoxybenzene, 2-4-methoxy-phenoxy ethylamine, 2-4-methoxy-phenoxy-ethylamine, 2-4-methoxyphenoxy ethan-1-amine, 2-4-methoxyphenoxy ethanamine, 2-4-methoxyphenoxy ethylamine, 2-4-methoxyphenoxy-1-ethanamine, ethanamine, 2-4-methoxyphenoxy, maybridge3_000601
COC1=CC=C(C=C1)OCCN
167.208
167.21
Technical
250mg
C9H13NO2
MFCD00052978
YXRCDWIZAGWUBL-UHFFFAOYSA-N
2-(4-methoxyphenoxy)ethanamine
1797793
Tech.
2-(4-Methoxyphenoxy)ethylamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS