Please login to your online account to display your discounted pricing

2-(4-methoxyphenoxy)ethanamine hydrochloride, 97%, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  BTB10107EA

 View more versions of this product

Catalog No. BTB10107EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 203.67
Quantity 10g
Chemical Name or Material 2-(4-methoxyphenoxy)ethanamine hydrochloride
CAS 98959-77-4
Molecular Formula C9H13NO2·ClH
Assay Percent Range 97%

Specifications

Chemical Name or Material 2-(4-methoxyphenoxy)ethanamine hydrochloride
Assay Percent Range 97%
Molecular Formula C9H13NO2·ClH
Formula Weight 203.67