Please login to your online account to display your discounted pricing

2-[4-(Methoxycarbonyl)-5-methyl-2-oxo-2,3-dihydro-1H-pyrrol-3-yl]acetic acid, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB09316DA

 View more versions of this product

Catalog No. BTB09316DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 213.19
Quantity 1g
Assay 97%
CAS Max % 100.0
Molecular Formula C9H11NO5
Packaging Amber Glass Bottle
Chemical Name or Material 2-[4-(Methoxycarbonyl)-5-methyl-2-oxo-2, 3-dihydro-1H-pyrrol-3-yl]acetic acid
CAS Min % 96.5
Assay Percent Range 97%
MDL Number MFCD00173859
CAS 77978-73-5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-[4-(Methoxycarbonyl)-5-methyl-2-oxo-2, 3-dihydro-1H-pyrrol-3-yl]acetic acid
CAS 77978-73-5
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H11NO5
Formula Weight 213.19
MDL Number MFCD00173859
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.