missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Methoxybenzylamino)pyridine, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC164150250

 View more versions of this product

Catalog No. AC164150250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-(4-Methoxybenzylamino)pyridine
Glass bottle
121°C to 126°C
97%
96.0
97%
MFCD00006252
SMJGJENXFSWIMQ-UHFFFAOYSA-N
N-[(4-methoxyphenyl)methyl]pyridin-2-amine
97452
Powder
Authentic
White to Yellow
25g
52818-63-0
100.0
C13H14N2O
n-4-methoxybenzyl pyridin-2-amine, 2-4-methoxybenzylamino pyridine, n-4-methoxyphenyl methyl pyridin-2-amine, p-anisyl 2-pyridyl amine, 4-methoxyphenyl methyl-2-pyridylamine, maybridge1_007212, 4-methoxybenzyl 2-pyridinylamine, 4-methoxy-benzyl-pyridin-2-yl-amine, 2-p-methoxybenzyl aminopyridine, n-4-methoxybenzyl-n-2-pyridyl amine
COC1=CC=C(C=C1)CNC2=CC=CC=N2
214.268
214.27
≥96.0%

Chemical Identifiers

52818-63-0
214.268
SMJGJENXFSWIMQ-UHFFFAOYSA-N
97452
COC1=CC=C(C=C1)CNC2=CC=CC=N2
C13H14N2O
MFCD00006252
n-4-methoxybenzyl pyridin-2-amine, 2-4-methoxybenzylamino pyridine, n-4-methoxyphenyl methyl pyridin-2-amine, p-anisyl 2-pyridyl amine, 4-methoxyphenyl methyl-2-pyridylamine, maybridge1_007212, 4-methoxybenzyl 2-pyridinylamine, 4-methoxy-benzyl-pyridin-2-yl-amine, 2-p-methoxybenzyl aminopyridine, n-4-methoxybenzyl-n-2-pyridyl amine
N-[(4-methoxyphenyl)methyl]pyridin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents