Please login to your online account to display your discounted pricing

2-(4-Methoxybenzylamino)pyridine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-(4-Methoxybenzylamino)pyridine
Name Note 97%
CAS 52818-63-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C13 H14 N2 O
Formula Weight 214.27
MDL Number MFCD00006252
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC164150050

  SDS
thermo fisher scientific
164150050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC164150250

  SDS
thermo fisher scientific
AC164150250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder
Formula Weight 214.27
Quantity 5g
Color White to yellow
Melting Point 121° to 126°C
CAS Max % 100.0
CAS Min % 96.0
CAS 52818-63-0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym N[(4-Methoxyphenyl)methyl]-2-pyridinamine
Chemical Name or Material 2-(4-Methoxybenzylamino)pyridine
Name Note 97%
MDL Number MFCD00006252
Molecular Formula C13 H14 N2 O
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.