missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Methoxybenzoyl)thiophene, TCI America™

$65.70 - $220.00

Chemical Identifiers

CAS 4160-63-8
Molecular Formula C12H10O2S
Molecular Weight (g/mol) 218.27
MDL Number MFCD00005440
InChI Key KYVBFEMQEUXVQB-UHFFFAOYSA-N
Synonym 2-4-methoxybenzoyl thiophene, 4-methoxyphenyl thiophen-2-yl methanone, 2-p-methoxybenzoyl thiophene, p-anisyl thiophen-2-yl ketone, 4-methoxyphenyl 2-thienyl ketone, methanone, 4-methoxyphenyl-2-thienyl, 4-methoxyphenyl 2-thienyl methanone, pubchem10833, acmc-1aikh, 2-thienyl p-anisyl ketone
PubChem CID 77812
IUPAC Name (4-methoxyphenyl)-thiophen-2-ylmethanone
SMILES COC1=CC=C(C=C1)C(=O)C2=CC=CS2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M23955G View Documents TCI America
M23955G
5g Each for $65.70
 
M239525G View Documents TCI America
M239525G
25g Each for $220.00
 
Specifications

Chemical Identifiers

4160-63-8
218.27
KYVBFEMQEUXVQB-UHFFFAOYSA-N
77812
COC1=CC=C(C=C1)C(=O)C2=CC=CS2
C12H10O2S
MFCD00005440
2-4-methoxybenzoyl thiophene, 4-methoxyphenyl thiophen-2-yl methanone, 2-p-methoxybenzoyl thiophene, p-anisyl thiophen-2-yl ketone, 4-methoxyphenyl 2-thienyl ketone, methanone, 4-methoxyphenyl-2-thienyl, 4-methoxyphenyl 2-thienyl methanone, pubchem10833, acmc-1aikh, 2-thienyl p-anisyl ketone
(4-methoxyphenyl)-thiophen-2-ylmethanone

Specifications

Undesignated
5g
C12H10O2S
2-4-methoxybenzoyl thiophene, 4-methoxyphenyl thiophen-2-yl methanone, 2-p-methoxybenzoyl thiophene, p-anisyl thiophen-2-yl ketone, 4-methoxyphenyl 2-thienyl ketone, methanone, 4-methoxyphenyl-2-thienyl, 4-methoxyphenyl 2-thienyl methanone, pubchem10833, acmc-1aikh, 2-thienyl p-anisyl ketone
COC1=CC=C(C=C1)C(=O)C2=CC=CS2
218.27
218.27
≥98.0% (GC)
74°C
4160-63-8
MFCD00005440
KYVBFEMQEUXVQB-UHFFFAOYSA-N
(4-methoxyphenyl)-thiophen-2-ylmethanone
77812
Crystal-Powder at 20°C
2-(4-Methoxybenzoyl)thiophene
Safety and Handling

Safety and Handling

TSCA : No