Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Methoxybenzoyl)thiophene, 97%

$118.99 - $489.49

Chemical Identifiers

Chemical Name or Material 2-(4-Methoxybenzoyl)thiophene
CAS 4160-63-8
Molecular Formula C12H10O2S
Formula Weight 218.27
MDL Number MFCD00005440
Beilstein 148560
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0551309

 
alfa aesar™
L0551309
10g Each for $118.99

AAL0551318

 
alfa aesar™
L0551318
50g Each for $489.49
Description & Specifications

Specifications

Formula Weight 218.27
Chemical Name or Material 2-(4-Methoxybenzoyl)thiophene
Assay 97%
Molecular Formula C12H10O2S
MDL Number MFCD00005440
CAS 4160-63-8
Beilstein 148560
Melting Point 71° to 75°C
Quantity 10g