missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
2,4-Mesitylenedisulfonyl Dichloride, TCI America™
Chemical Identifiers
CAS |
68985-08-0 |
Molecular Formula |
C9H10Cl2O4S2 |
Molecular Weight (g/mol) |
317.195 |
MDL Number |
MFCD00009796 |
InChI Key |
LNXKRGBQKLXFTB-UHFFFAOYSA-N |
Synonym |
2,4,6-Trimethyl-1,3-benzenedisulfonyl Dichloride |
PubChem CID |
3620081 |
IUPAC Name |
2,4,6-trimethylbenzene-1,3-disulfonyl chloride |
SMILES |
CC1=CC(=C(C(=C1S(=O)(=O)Cl)C)S(=O)(=O)Cl)C |
View More Specs
Products
2
Specifications
Chemical Identifiers
|
68985-08-0 |
|
317.195 |
|
LNXKRGBQKLXFTB-UHFFFAOYSA-N |
|
3620081 |
|
CC1=CC(=C(C(=C1S(=O)(=O)Cl)C)S(=O)(=O)Cl)C |
|
C9H10Cl2O4S2 |
|
MFCD00009796 |
|
2,4,6-Trimethyl-1,3-benzenedisulfonyl Dichloride |
|
2,4,6-trimethylbenzene-1,3-disulfonyl chloride |
Specifications
|
White |
|
5g |
|
C9H10Cl2O4S2 |
|
3261 |
|
LNXKRGBQKLXFTB-UHFFFAOYSA-N |
|
2,4,6-trimethylbenzene-1,3-disulfonyl chloride |
|
3620081 |
|
Crystal-Powder at 20°C |
|
2,4-Mesitylenedisulfonyl Dichloride |
|
125°C |
|
68985-08-0 |
|
MFCD00009796 |
|
2,4,6-Trimethyl-1,3-benzenedisulfonyl Dichloride |
|
CC1=CC(=C(C(=C1S(=O)(=O)Cl)C)S(=O)(=O)Cl)C |
|
317.195 |
|
317.20 |
|
≥98.0% (T) |
Safety and Handling