Learn More
2,4-Lutidine, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 125181000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2, 4-Lutidine | |
108-47-4 | |
100.0 | |
159.0°C | |
98% min. (GC) | |
C7H9N | |
MFCD00006337 | |
, 244 | |
0.92 | |
Solubility in water: 15g/100 g (20°C). Other solubilities: soluble in alcohol,ether,acetone and,benzene | |
CC1=CC=NC(C)=C1 | |
107.16 | |
107.15 |
98+% | |
98.0 | |
0.9200g/mL | |
47°C | |
Glass bottle | |
1.4990 to 1.5010 | |
100mL | |
06,224 | |
2,4-lutidine, pyridine, 2,4-dimethyl, alpha,gamma-dimethylpyridine, 2,4-lutidene, unii-83903uj0ww, 2,4-dimethyl-pyridine, .alpha.,.gamma.-dimethylpyridine, 2,4-dimethyl pyridine, 2.4-lutidine, 2,4 lutidine | |
JYYNAJVZFGKDEQ-UHFFFAOYSA-N | |
2,4-dimethylpyridine | |
7936 | |
98+% |
Chemical Identifiers
108-47-4 | |
107.16 | |
JYYNAJVZFGKDEQ-UHFFFAOYSA-N | |
7936 | |
CC1=CC=NC(C)=C1 |
C7H9N | |
MFCD00006337 | |
2,4-lutidine, pyridine, 2,4-dimethyl, alpha,gamma-dimethylpyridine, 2,4-lutidene, unii-83903uj0ww, 2,4-dimethyl-pyridine, .alpha.,.gamma.-dimethylpyridine, 2,4-dimethyl pyridine, 2.4-lutidine, 2,4 lutidine | |
2,4-dimethylpyridine |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Toxic if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at
GHS Signal Word: Danger
EINECSNumber : 203-586-8
RTECSNumber : OK9400000
TSCA : TSCA
RUO â Research Use Only