Learn More
2-(4-Isocyanatophenyl)pyrimidine, 90%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC56206DA
Specifications
2-(4-Isocyanatophenyl)pyrimidine | |
89.5 | |
90% | |
C11H7N3O | |
1g | |
VGGNOZLRRMBBQL-UHFFFAOYSA-N | |
2-(4-isocyanatophenyl)pyrimidine | |
24229558 | |
90% |
216059-84-6 | |
100.0 | |
Amber Glass Bottle | |
MFCD09702380 | |
2-4-isocyanatophenyl pyrimidine, 4-pyrimidin-2-ylbenzenisocyanate, 4-pyrimidin-2-yl phenylisocyanate, pyrimidine,2-4-isocyanatophenyl, 4-pyrimidin-2-yl phenyl isocyanate | |
O=C=NC1=CC=C(C=C1)C1=NC=CC=N1 | |
197.20 | |
197.2 |
Chemical Identifiers
216059-84-6 | |
197.20 | |
VGGNOZLRRMBBQL-UHFFFAOYSA-N | |
24229558 | |
O=C=NC1=CC=C(C=C1)C1=NC=CC=N1 |
C11H7N3O | |
MFCD09702380 | |
2-4-isocyanatophenyl pyrimidine, 4-pyrimidin-2-ylbenzenisocyanate, 4-pyrimidin-2-yl phenylisocyanate, pyrimidine,2-4-isocyanatophenyl, 4-pyrimidin-2-yl phenyl isocyanate | |
2-(4-isocyanatophenyl)pyrimidine |
Safety and Handling
GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger