Please login to your online account to display your discounted pricing

2-(4-Isobutylphenyl)propanenitrile, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SEW06499DA

 View more versions of this product

Catalog No. SEW06499DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 187.28
Quantity 1g
Assay 97%
CAS Min % 97.0
Assay Percent Range 97%
Linear Formula (CH3)2CHCH2C6H4CH(CN)CH3
MDL Number MFCD02180706
CAS 58609-73-7
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Molecular Formula C13H17N
Packaging Amber Glass Bottle
Chemical Name or Material 2-(4-Isobutylphenyl)propanenitrile
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-(4-Isobutylphenyl)propanenitrile
CAS 58609-73-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H17N
Linear Formula (CH3)2CHCH2C6H4CH(CN)CH3
Formula Weight 187.28
MDL Number MFCD02180706
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

SDS & Safety Documents