missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Iodophenyl)-1-phenylbenzimidazole, TCI America™

$172.64 - $594.31

Chemical Identifiers

CAS 760212-42-8
Molecular Formula C19H13IN2
Molecular Weight (g/mol) 396.231
InChI Key AZRBPNMWQFEAJW-UHFFFAOYSA-N
PubChem CID 23094068
IUPAC Name 2-(4-iodophenyl)-1-phenylbenzimidazole
SMILES C1=CC=C(C=C1)N2C3=CC=CC=C3N=C2C4=CC=C(C=C4)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I09831G View Documents TCI America
I09831G
1g Each for $594.31
 
I0983200MG View Documents TCI America
I0983200MG
200mg Each for $172.64
 
Specifications

Chemical Identifiers

760212-42-8
396.231
23094068
C1=CC=C(C=C1)N2C3=CC=CC=C3N=C2C4=CC=C(C=C4)I
C19H13IN2
AZRBPNMWQFEAJW-UHFFFAOYSA-N
2-(4-iodophenyl)-1-phenylbenzimidazole

Specifications

White-Yellow
1g
C19H13IN2
C1=CC=C(C=C1)N2C3=CC=CC=C3N=C2C4=CC=C(C=C4)I
396.231
396.23
≥98.0% (HPLC,T)
180°C
760212-42-8
AZRBPNMWQFEAJW-UHFFFAOYSA-N
2-(4-iodophenyl)-1-phenylbenzimidazole
23094068
Crystal-Powder at 20°C
2-(4-Iodophenyl)-1-phenylbenzimidazole
Safety and Handling

Safety and Handling

TSCA : No