missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(4-Iodophenyl)-1-phenylbenzimidazole 98.0+%, TCI America™
Supplier: TCI America I09831G
Specifications
2-(4-Iodophenyl)-1-phenylbenzimidazole | |
180°C | |
C19H13IN2 | |
1 g | |
IC1=CC=C(C=C1)C1=NC2=CC=CC=C2N1C1=CC=CC=C1 | |
396.23 | |
396.23 | |
Crystalline Powder |
760212-42-8 | |
White-Yellow | |
MFCD29089366 | |
AZRBPNMWQFEAJW-UHFFFAOYSA-N | |
2-(4-iodophenyl)-1-phenyl-1H-1,3-benzodiazole | |
23094068 | |
≥98.0% (HPLC,T) |
Chemical Identifiers
760212-42-8 | |
396.23 | |
AZRBPNMWQFEAJW-UHFFFAOYSA-N | |
2-(4-iodophenyl)-1-phenyl-1H-1,3-benzodiazole |
C19H13IN2 | |
MFCD29089366 | |
23094068 | |
IC1=CC=C(C=C1)C1=NC2=CC=CC=C2N1C1=CC=CC=C1 |
Safety and Handling
TSCA : No