Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Hydroxyphenylazo)benzoic acid, Ultrapure MALDI Matrix

For use as a matrix in MALDI mass spectrometry analysis

Chemical Identifiers

Chemical Name or Material 2-(4-Hydroxyphenylazo)benzoic acid
CAS 1634-82-8
Name Note Ultrapure MALDI Matrix
Formula Weight 242.23
MDL Number MFCD00002428
Beilstein 1842696
Synonym 2-(4-Hydroxyphenylazo) benzoic acid; HABA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67769EXK

  SDS
alfa aesar™
J67769EXK
5 x 10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 204° to 208°C
Formula Weight 242.23
MDL Number MFCD00002428
TSCA TSCA
Synonym 2-(4-Hydroxyphenylazo) benzoic acid; HABA
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-(4-Hydroxyphenylazo)benzoic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 1634-82-8
Beilstein 1842696
Name Note Ultrapure MALDI Matrix
Health Hazard 1 Warning
Quantity 5 x 10mg
Color Yellow to orange
Physical Form Powder
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.